Learn More
Shimadzu Scientific Instruments Sample Vials with Caps and Septa, 23mm Diameter, 30 per Pack
Catalog No. NC2889542
Encompass
Use with Rack LL and SIL-10AF or AP only. Not compatible with the SIL-10A. Can be used with FRC-40/LH-40 Rack Kit D25.
Catalog No. NC2889542 Supplier Shimadzu Scientific Instruments Supplier No. 2282128092
Use with Rack LL and SIL-10AF or AP only. Not compatible with the SIL-10A. Can be used with FRC-40/LH-40 Rack Kit D25.