Learn More
Thermo Scientific Valve Cartridge Vf-P20 Ncs Nc 1 Ea
Supplier:  Thermo Scientific 6041.2301
Encompass
Thermal regulator valve ™ cartridge for high performance liquid chromatography systems with 20 UL sample loop provides switching of mobile phases for gradient elution of analytes constructed of chemically inert materials enables precise flow control for optimal separation and quantification of analytes.
Catalog No. 50-134-7893