Learn More
Chemglass Life Sciences Valve, Chem-Vac, Hi-Vac, 180°, 0-4mm, Viton O-Ring
SDP
Catalog No. CG92701
Encompass_Preferred
Valve, Chem-Vac, Hi-Vac, 180¿, 0-4mm, Viton O-Ring. CHEM-VAC valve with two sidearms sealed to the barrel at a 180¿ angle to one another and on the same plane. Valve has TFE plug with three o-rings, the tip o-ring making the primary seal on the precision formed tapered seat. O-rings on the non-rotating shaft form an excellent dynamic seal between the TFE plug and glass barrel. See CG-935 for replacement plug, CG-537 for o-ring replacement kit, and CG-904 for glass portion only.
Catalog No. CG92701 Supplier Chemglass Life Sciences Supplier No. CG92701
Valve, Chem-Vac, Hi-Vac, 180¿, 0-4mm, Viton O-Ring. CHEM-VAC valve with two sidearms sealed to the barrel at a 180¿ angle to one another and on the same plane. Valve has TFE plug with three o-rings, the tip o-ring making the primary seal on the precision formed tapered seat. O-rings on the non-rotating shaft form an excellent dynamic seal between the TFE plug and glass barrel. See CG-935 for replacement plug, CG-537 for o-ring replacement kit, and CG-904 for glass portion only.