Learn More
Chemglass Life Sciences Valve, Chem-Vac, Hi-Vac, 90°, 0-12mm Bore, Viton O-Ring
SDP
Catalog No. CG92503
Encompass_Preferred
Valve, Chem-Vac, Hi-Vac, 90¿, 0-12mm Bore, Viton O-Ring. CHEM-VAC valve with two sidearms sealed to the barrel at a 90¿ angle to one another. Valve has a TFE plug with three o-rings, the tip o-ring making the primary seal on the precision formed tapered seat. O-rings on the non-rotating shaft form an excellent dynamic seal between the TFE plug and glass barrel. See CG-935 for replacement plug, CG-537 for o-ring replacement kit, and CG-903 for glass portion only.
Catalog No. CG92503 Supplier Chemglass Life Sciences Supplier No. CG92503
Valve, Chem-Vac, Hi-Vac, 90¿, 0-12mm Bore, Viton O-Ring. CHEM-VAC valve with two sidearms sealed to the barrel at a 90¿ angle to one another. Valve has a TFE plug with three o-rings, the tip o-ring making the primary seal on the precision formed tapered seat. O-rings on the non-rotating shaft form an excellent dynamic seal between the TFE plug and glass barrel. See CG-935 for replacement plug, CG-537 for o-ring replacement kit, and CG-903 for glass portion only.