Learn More
Chemglass Life Sciences Valve, Chem-Vac, Chem-Cap, Hi-Vac, 180°, 0-8mm Bore
SDP
Catalog No. CG95302
Encompass_Preferred
Valve, Chem-Vac, Chem-Cap, Hi-Vac, 180, 0-8mm Bore. CHEM-VAC CHEM-CAP valve with two sidearms sealed to the barrel at a 180 angle to one another and on the same plane. See CG-961 for replacement CHEM-CAP plug and CG-904 for glass portion only.
Catalog No. CG95302 Supplier Chemglass Life Sciences Supplier No. CG95302
Valve, Chem-Vac, Chem-Cap, Hi-Vac, 180, 0-8mm Bore. CHEM-VAC CHEM-CAP valve with two sidearms sealed to the barrel at a 180 angle to one another and on the same plane. See CG-961 for replacement CHEM-CAP plug and CG-904 for glass portion only.