Learn More
Chemglass Life Sciences Valve, Chem-Vac, Chem-Cap, Hi-Vac, Three-Way, 0-4mm Bore
SDP
Catalog No. CG95701
Encompass_Preferred
Valve, Chem-Vac, Chem-Cap, Hi-Vac, Three-Way, 0-4mm Bore. CHEM-VAC CHEM-CAP valve with three-way configuration for use in construction of manifolds, adapters, etc. See CG-961 for replacement CHEM-CAP plug and CG-906 for glass portion only.
Catalog No. CG95701 Supplier Chemglass Life Sciences Supplier No. CG95701
Valve, Chem-Vac, Chem-Cap, Hi-Vac, Three-Way, 0-4mm Bore. CHEM-VAC CHEM-CAP valve with three-way configuration for use in construction of manifolds, adapters, etc. See CG-961 for replacement CHEM-CAP plug and CG-906 for glass portion only.