Learn More
Chemglass Life Sciences Valve, Chem-Vac, Chem-Cap, Hi-Vac, 90°, 0-4mm Bore
SDP
Catalog No. CG95101
Encompass_Preferred
Valve, Chem-Vac, Chem-Cap, Hi-Vac, 90, 0-4mm Bore. CHEM-VAC CHEM-CAP valve with two sidearms sealed to the barrel at a 90 angle to one another. See CG-961 for replacement CHEM-CAP plug and CG-903 for glass portion only.
Catalog No. CG95101 Supplier Chemglass Life Sciences Supplier No. CG95101
Valve, Chem-Vac, Chem-Cap, Hi-Vac, 90, 0-4mm Bore. CHEM-VAC CHEM-CAP valve with two sidearms sealed to the barrel at a 90 angle to one another. See CG-961 for replacement CHEM-CAP plug and CG-903 for glass portion only.