Learn More
Chemglass Life Sciences Valve, Chem-Vac, Chem-Cap, Hi-Vac, Metering, 1-Arm, 0-4mm Bore
SDP
Catalog No. CG96201
Encompass_Preferred
Valve, Chem-Vac, Chem-Cap, Hi-Vac, Metering, 1-Arm, 0-4mm Bore. CHEM-VAC CHEM-CAP metering valve has an extended tip on the plug to allow semi-needle valve control of gases and liquids. Valve has single sidearm sealed at a 90 angle to the barrel. This inexpensive configuration allows for the lower portion of the barrel to be sealed directly to the apparatus being constructed, making for more compact, stronger assemblies. See CG-971 for replacement CHEM-CAP metering plug and CG-902 for glass portion only.
Catalog No. CG96201 Supplier Chemglass Life Sciences Supplier No. CG96201
Valve, Chem-Vac, Chem-Cap, Hi-Vac, Metering, 1-Arm, 0-4mm Bore. CHEM-VAC CHEM-CAP metering valve has an extended tip on the plug to allow semi-needle valve control of gases and liquids. Valve has single sidearm sealed at a 90 angle to the barrel. This inexpensive configuration allows for the lower portion of the barrel to be sealed directly to the apparatus being constructed, making for more compact, stronger assemblies. See CG-971 for replacement CHEM-CAP metering plug and CG-902 for glass portion only.