Learn More
Medchemexpress LLC HY-12471 1mg Medchemexpress, VLX1570 CAS:1431280-51-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY124711MG
Encompass_Preferred
Medchemexpress, HY-12471 1mg VLX1570 CAS:1431280-51-1 VLX1570 is a competitive inhibitor of proteasome deubiquitinases (DUBs) with an IC50 of approximate 10 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8883