Learn More
TARGETMOL CHEMICALS INC VLX1570 5MG
SDP
Catalog No. 502579976
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. VLX1570 is a competitive inhibitor of proteasome DUB activity with IC50 ranging from 4.2 uM to 8.6 uM. It has potent inhibition for USP14. purity: 99%
Catalog No. 50-257-9976 Supplier TARGETMOL CHEMICALS INC Supplier No. T40675MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. VLX1570 is a competitive inhibitor of proteasome DUB activity with IC50 ranging from 4.2 uM to 8.6 uM. It has potent inhibition for USP14. purity: 99%