Learn More
Selleck Chemical LLC VLX1570 5mg 1431280-51-1
SDP
Catalog No. 501365904
Encompass_Preferred
VLX1570 is a competitive inhibitor of proteasome DUB activity, with an IC50 of ~10??M in vitro. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5904 Supplier Selleck Chemical LLC Supplier No. S82885MG
Add to Cart
Add to Cart
VLX1570 is a competitive inhibitor of proteasome DUB activity, with an IC50 of ~10??M in vitro. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.