Learn More
TARGETMOL CHEMICALS INC VLX600 25MG
SDP
Catalog No. 502579064
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. VLX600 is an iron-chelating oxidative phosphorylation (OXPHOS) inhibitor is a cell-permeable anticancer agent. It acts by reducing mitochondrial oxidative phosphorylation in tumor cells. purity: 100%
Catalog No. 50-257-9064 Supplier TARGETMOL CHEMICALS INC Supplier No. T850025MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. VLX600 is an iron-chelating oxidative phosphorylation (OXPHOS) inhibitor is a cell-permeable anticancer agent. It acts by reducing mitochondrial oxidative phosphorylation in tumor cells. purity: 100%