Learn More
Medchemexpress LLC Vlx600 | 327031-55-0 | MFCD01305422 | 99.8% | 317.35 | C17H15N7 | 100MG
SDP
Catalog No. 5000235125
Encompass_Preferred
VLX600 is a small-molecule oxidative phosphorylation (OxPhos) inhibitor and iron chelator used in cancer and mitochondrial metabolism research. It impairs mitochondrial respiration, induces autophagy, and shows selective cytotoxicity in quiescent or nutrient-starved tumor cells. Molecular formula C17H15N7; molecular weight 317.35; CAS 327031-55-0.
  • Inhibits mitochondrial oxidative phosphorylation.
  • Functions as an iron chelator.
  • Induces autophagy and promotes tumor cell death under nutrient limitation.
  • Useful as a reference compound for cancer metabolism studies.
  • Crystalline solid with reported DMSO solubility around 25 mg/mL.
  • Recommended storage at -20°C as a powder.
Catalog No. 50-002-35125 Supplier Medchemexpress LLC Supplier No. HY12406100MG
VLX600 is a small-molecule oxidative phosphorylation (OxPhos) inhibitor and iron chelator used in cancer and mitochondrial metabolism research. It impairs mitochondrial respiration, induces autophagy, and shows selective cytotoxicity in quiescent or nutrient-starved tumor cells. Molecular formula C17H15N7; molecular weight 317.35; CAS 327031-55-0.
  • Inhibits mitochondrial oxidative phosphorylation.
  • Functions as an iron chelator.
  • Induces autophagy and promotes tumor cell death under nutrient limitation.
  • Useful as a reference compound for cancer metabolism studies.
  • Crystalline solid with reported DMSO solubility around 25 mg/mL.
  • Recommended storage at -20°C as a powder.