Learn More
Sigma Aldrich Fine Chemicals Biosciences VM20 Vacuum Manifold for p
Catalog No. 502901758
Encompass
The VM20 Vacuum Manifold is designed to provide increased efficiency and performance when used with many of Sigmas column-based nucleic acid purification systems. The manifold is capable of processing 20 samples simultaneously and includes a stepped design for easy access to every column. Dual vacuum connection ports allow for multiple configurations for waste collection. Waste can be collected in the manifold body or through use of a threaded drain port diverted to any auxiliary reservoir providing virtually unlimited capacity. The manifold includes chemically resistant Endura-Luer stopcocks that will not crack or break with repeated use.
Catalog No. 50-290-1758 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. VM201SET
The VM20 Vacuum Manifold is designed to provide increased efficiency and performance when used with many of Sigmas column-based nucleic acid purification systems. The manifold is capable of processing 20 samples simultaneously and includes a stepped design for easy access to every column. Dual vacuum connection ports allow for multiple configurations for waste collection. Waste can be collected in the manifold body or through use of a threaded drain port diverted to any auxiliary reservoir providing virtually unlimited capacity. The manifold includes chemically resistant Endura-Luer stopcocks that will not crack or break with repeated use.