Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC VMIP-II (1-21) TFA | 5 MG
SDP

Catalog No. 5000398531
Encompass_Preferred

vMIP-II (1-21) (NT21MP) TFA is an inhibitor of CXCR4. It interacts broadly with CC and CXC chemokine receptors, inhibiting CXCR4 by competing with 125I-SDF-1R for binding sites (IC50=190 nM). This product is for research use only and has not been fully validated for medical applications.

  • Molecular formula: C108H169N33O27S.xC2HF3O2
  • Sequence: Leu-Gly-Ala-Ser-Trp-His-Arg-Pro-Asp-Lys-Cys-Cys-Leu-Gly-Tyr-Gln-Lys-Arg-Pro-Leu-Pro
  • Target: CXCR
  • Pathway: GPCR/G protein; immunology/inflammation

Catalog No. 50-003-98531 Supplier Medchemexpress LLC Supplier No. HYP4109A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

vMIP-II (1-21) (NT21MP) TFA is an inhibitor of CXCR4. It interacts broadly with CC and CXC chemokine receptors, inhibiting CXCR4 by competing with 125I-SDF-1R for binding sites (IC50=190 nM). This product is for research use only and has not been fully validated for medical applications.

  • Molecular formula: C108H169N33O27S.xC2HF3O2
  • Sequence: Leu-Gly-Ala-Ser-Trp-His-Arg-Pro-Asp-Lys-Cys-Cys-Leu-Gly-Tyr-Gln-Lys-Arg-Pro-Leu-Pro
  • Target: CXCR
  • Pathway: GPCR/G protein; immunology/inflammation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.