Learn More
Cayman Chemical Vinpocetine, Purity: ≥98%, CAS: 42971-09-5, 50 mg
SDP
Catalog No. 502983163
Encompass_Preferred
An inhibitor of PDE1 that has diverse cerebral and neurological effects in vivo; also directly inhibits IKKβ (IC50 = 17 µM in a cell-free system) and blocks TNF-α- and LPS-mediated activation of NF-κB in cells and in vivo
Catalog No. 50-298-3163 Supplier Cayman Chemical Supplier No. 1957850 MG
An inhibitor of PDE1 that has diverse cerebral and neurological effects in vivo; also directly inhibits IKKβ (IC50 = 17 µM in a cell-free system) and blocks TNF-α- and LPS-mediated activation of NF-κB in cells and in vivo