Learn More
Medchemexpress LLC VNT-101 | 2894060-67-2 | 99.5% | 456.41 | 5 MG
SDP
Catalog No. 5000386405
Encompass_Preferred
VNT-101 is an orally active influenza A (IAV) inhibitor that disrupts NP-NP PPI to block NP oligomerization and destabilize the viral ribonucleoprotein (RNP) complex. It exhibits potent antiviral activity across multiple influenza A subtypes and robust in vivo antiviral efficacy in mice infected with lethal H1N1 virus. It can be used for the study of influenza A infection.
  • Orally active influenza A (IAV) inhibitor.
  • Disrupts NP-NP PPI to block NP oligomerization.
  • Destabilizes the viral ribonucleoprotein (RNP) complex.
  • Potent antiviral activity across multiple influenza A subtypes.
  • Exhibits EC50 values of 4-5 nM in cellular cytopathic effect (CPE) assay, 4-8 nM in neuraminidase (NA) assay, and 21-45 nM in RNP assay.
  • Demonstrates robust in vivo antiviral efficacy in mice infected with lethal H1N1 virus.
Catalog No. 50-003-86405 Supplier Medchemexpress LLC Supplier No. HY1754695MG
VNT-101 is an orally active influenza A (IAV) inhibitor that disrupts NP-NP PPI to block NP oligomerization and destabilize the viral ribonucleoprotein (RNP) complex. It exhibits potent antiviral activity across multiple influenza A subtypes and robust in vivo antiviral efficacy in mice infected with lethal H1N1 virus. It can be used for the study of influenza A infection.
  • Orally active influenza A (IAV) inhibitor.
  • Disrupts NP-NP PPI to block NP oligomerization.
  • Destabilizes the viral ribonucleoprotein (RNP) complex.
  • Potent antiviral activity across multiple influenza A subtypes.
  • Exhibits EC50 values of 4-5 nM in cellular cytopathic effect (CPE) assay, 4-8 nM in neuraminidase (NA) assay, and 21-45 nM in RNP assay.
  • Demonstrates robust in vivo antiviral efficacy in mice infected with lethal H1N1 virus.