Learn More
Uline Uline Industrial Vinyl Safety Tape - 1 x 36 yds | Yellow | 1 EA
Catalog No. NC2246019
Encompass_Preferred
This item must be ordered in multiples of 3 per supplier requirements
Industrial 6 mil vinyl with rubber adhesive - unlaminated general purpose, Non-reflective, Adheres to clean dry smooth non-porous surfaces, Not recommended for outdoor use, Use solvent to remove adhesive
This item is not returnable. View return policy
Catalog No. NC2246019 Supplier Uline Supplier No. S7190
This item must be ordered in multiples of 3 per supplier requirements
Industrial 6 mil vinyl with rubber adhesive - unlaminated general purpose, Non-reflective, Adheres to clean dry smooth non-porous surfaces, Not recommended for outdoor use, Use solvent to remove adhesive