Learn More
Apexbio Technology LLC VO-Ohpic trihydrate 476310-60-8 10mg
SDP
Catalog No. 5000095471
Encompass_Preferred
VO-Ohpic trihydrate (CAS 476310-60-8) is a highly selective small-molecule inhibitor of the lipid phosphatase PTEN (phosphatase and tensin homolog) with an in vitro IC50 of 35 nM VO-Ohpic shows marked selectivity for PTEN over other phosphatases including CBPs SopB myotubularin SAC1 and PTP- In cell-based assays VO-Ohpic induces dose-dependent phosphorylation of Akt at Ser473 and Thr308 saturating at 75 nM and promotes Akt functional activation as indicated by decreased FoxO3a transcriptional activity In vivo VO-Ohpic administration in MDA PCa-2b tumor xenograft mice results in reduced tumor growth and increased survival This compound serves as a valuable tool for investigating PTEN function and its modulation in insulin signaling glucose uptake and cancer research
Catalog No. 50-000-95471 Supplier Apexbio Technology LLC Supplier No. A392310
VO-Ohpic trihydrate (CAS 476310-60-8) is a highly selective small-molecule inhibitor of the lipid phosphatase PTEN (phosphatase and tensin homolog) with an in vitro IC50 of 35 nM VO-Ohpic shows marked selectivity for PTEN over other phosphatases including CBPs SopB myotubularin SAC1 and PTP- In cell-based assays VO-Ohpic induces dose-dependent phosphorylation of Akt at Ser473 and Thr308 saturating at 75 nM and promotes Akt functional activation as indicated by decreased FoxO3a transcriptional activity In vivo VO-Ohpic administration in MDA PCa-2b tumor xenograft mice results in reduced tumor growth and increased survival This compound serves as a valuable tool for investigating PTEN function and its modulation in insulin signaling glucose uptake and cancer research