Learn More
Medchemexpress LLC VO-Ohpic trihydrate | 476310-60-8 | 98.0% | 100 MG
SDP
Catalog No. 5000339800
Encompass_Preferred
VO-Ohpic trihydrate is a highly potent inhibitor of PTEN with an IC50 of 46±10 nM.
  • Highly potent inhibitor of PTEN with an IC50 of 46±10 nM.
  • Significantly inhibits PTEN activity in low nanomolar concentrations.
  • An encouragingly specific and potent PTEN inhibitor.
  • The most potent inhibitor (IC50=35 nM) of the PTEN lipid phosphatase activity.
Catalog No. 50-003-39800 Supplier Medchemexpress LLC Supplier No. HY13074100MG
VO-Ohpic trihydrate is a highly potent inhibitor of PTEN with an IC50 of 46±10 nM.
  • Highly potent inhibitor of PTEN with an IC50 of 46±10 nM.
  • Significantly inhibits PTEN activity in low nanomolar concentrations.
  • An encouragingly specific and potent PTEN inhibitor.
  • The most potent inhibitor (IC50=35 nM) of the PTEN lipid phosphatase activity.