Learn More
Medchemexpress LLC VO-OHpic trihydrate | 476310-60-8 | 98.0% | C12H16N2O11V | 10MG
SDP
Catalog No. 5000261992
Encompass_Preferred
VO-Ohpic trihydrate is a vanadium-containing small-molecule inhibitor of PTEN used in biochemical and cell-based research to probe PI3K/AKT signaling. It reversibly inhibits PTEN with low-nanomolar potency and is supplied as the trihydrate form to aid stability and handling.
  • Potent PTEN inhibition (IC50 ≈ 46 ± 10 nM).
  • Vanadium-containing complex for targeted phosphatase interaction.
  • Suitable for biochemical and cell-based assays probing PI3K/AKT signaling.
  • Trihydrate form to improve compound stability during storage and handling.
  • Available in small, research-grade quantities for laboratory use.
Catalog No. 50-002-61992 Supplier Medchemexpress LLC Supplier No. HY1307410MG
VO-Ohpic trihydrate is a vanadium-containing small-molecule inhibitor of PTEN used in biochemical and cell-based research to probe PI3K/AKT signaling. It reversibly inhibits PTEN with low-nanomolar potency and is supplied as the trihydrate form to aid stability and handling.
  • Potent PTEN inhibition (IC50 ≈ 46 ± 10 nM).
  • Vanadium-containing complex for targeted phosphatase interaction.
  • Suitable for biochemical and cell-based assays probing PI3K/AKT signaling.
  • Trihydrate form to improve compound stability during storage and handling.
  • Available in small, research-grade quantities for laboratory use.