Learn More
Medchemexpress LLC HY-13074 5mg Medchemexpress, VO-Ohpic (trihydrate) CAS:476310-60-8 Purity:>98%
SDP
Catalog No. 502030227
Encompass_Preferred
Medchemexpress, HY-13074 5mg VO-Ohpic (trihydrate) CAS:476310-60-8 VO-Ohpic trihydrate is a highly potent inhibitor of PTEN with an IC50 of 46±10 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0227 Supplier Medchemexpress LLC Supplier No. HY130745MG
Medchemexpress, HY-13074 5mg VO-Ohpic (trihydrate) CAS:476310-60-8 VO-Ohpic trihydrate is a highly potent inhibitor of PTEN with an IC50 of 46±10 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.