Learn More
Scientific Specialties Service Inc Clear Borosilicate Screw Thread Vials with Sci/Spec Teflon Lined Caps
SDP
Catalog No. NC1709845
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
Clear Borosilicate Screw Thread Vials with Sci/Spec Teflon Lined Caps, vial size: 27mm x 57mm, cap size: 24-400
Catalog No. NC1709845 Supplier Scientific Specialties Service Inc Supplier No. B75520
This item has a minimum qty of 2 per supplier requirements.
Clear Borosilicate Screw Thread Vials with Sci/Spec Teflon Lined Caps, vial size: 27mm x 57mm, cap size: 24-400