Learn More
Cayman Chemical Voacamine, Purity: ≥98%, CAS: 3371-85-5, 10 mg
SDP
Catalog No. 502934925
Encompass_Preferred
An alkaloid with diverse biological activities; a CB1 receptor antagonist (IC50 = 41 nM); time- and dose-dependently decreases survival of U2OS and U2OS/DX osteosarcoma cells and induces autophagy, but not apoptosis, at 1-10 µg/ml; inhibits growth of P. falciparum in vitro and reduces parasitemia in mice infected with P. yoelii when administered at doses of 2.5, 5, and 10 mg/kg
Catalog No. 50-293-4925 Supplier Cayman Chemical Supplier No. 2520410 MG
An alkaloid with diverse biological activities; a CB1 receptor antagonist (IC50 = 41 nM); time- and dose-dependently decreases survival of U2OS and U2OS/DX osteosarcoma cells and induces autophagy, but not apoptosis, at 1-10 µg/ml; inhibits growth of P. falciparum in vitro and reduces parasitemia in mice infected with P. yoelii when administered at doses of 2.5, 5, and 10 mg/kg