Learn More
Cayman Chemical VoacngarIn 10mg
SDP
Catalog No. 503015749
Encompass_Preferred
A cytotoxic indole alkaloid; cytotoxic to HepG2, A375, MDA-MB-231, SH-SY5Y, and CT26 cells (IC50s = 20 UG/ml); cytostatic to wild-type S. cerevisiae and cytotoxic to Δrad3-e5 and Δrad6-1 mutant S. cerevisiae
Catalog No. 50-301-5749 Supplier Cayman Chemical Supplier No. 2546810 MG
A cytotoxic indole alkaloid; cytotoxic to HepG2, A375, MDA-MB-231, SH-SY5Y, and CT26 cells (IC50s = 20 UG/ml); cytostatic to wild-type S. cerevisiae and cytotoxic to Δrad3-e5 and Δrad6-1 mutant S. cerevisiae