Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VOC Mixture, 200μg/mL in Methanol, Ultra Scientific

Catalog No. USDWM8261
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USDWM8261 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USDWM8261 Supplier Agilent Technologies Supplier No. DWM8261
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Specifications

CAS Max % 98.65, 0.02, 0.02, 0.02, 0.02
CAS 75-27-4, 75-25-2, 56-23-5, 67-66-3, 124-48-1
Physical Form Liquid
Quantity 1 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.