Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ VOC-Pak™ Polisher

Catalog No. V0CPAK001 Shop All MilliporeSigma Products
Change view
Click to view available options
For Use With (Equipment):
Direct-Q™ UV, Direct-Q™, Milli-Q™ Direct, Milli-Q™ Reference, Simplicity™ UV, Simplicity™, Synergy™ UV, Synergy™
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
V0CPAK001 Direct-Q™ UV, Direct-Q™, Milli-Q™ Direct, Milli-Q™ Reference, Simplicity™ UV, Simplicity™, Synergy™ UV, Synergy™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. V0CPAK001 Supplier MilliporeSigma™ Supplier No. V0CPAK001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MilliporeSigma VOC-Pak™ Polisher - Production of volatile organic compound-free water at the point of dispense.

  • Validated to produce 300 L of VOC-free ultrapure water.
  • Easy maintenance: the polisher is easily installed and replaced.
  • Bottom-tip is redesigned for a secured installation of the protection bell.
  • e-Sure tag for RFID connection with Q-POD™ dispenser enables full traceability (data management) and automatic consumable status monitoring on the POD's touchscreen interface.
  • Certificate of Quality available online.
  • Provides a warranty of water suitable for VOC analysis: blanks and standards preparation; glassware cleaning; and sample dissolution (if VOCs are analyzed in a matrix other than water i.e., in soil).
TRUSTED_SUSTAINABILITY

Specifications

Description Volatile Organic Compound-Free Water
For Use With (Equipment) Direct-Q™ UV, Direct-Q™, Milli-Q™ Direct, Milli-Q™ Reference, Simplicity™ UV, Simplicity™, Synergy™ UV, Synergy™
Type Polisher
Flow Rate ≤2.0 L/min.
Material Polypropylene
Resistivity >18.0 MΩ-cm (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.