Learn More
Medchemexpress LLC Vocacapsaicin hydrochloride | 1931116-92-5 | 95.7% | 496.08 | 1 MG
SDP
Catalog No. 5000351790
Encompass_Preferred
Vocacapsaicin hydrochloride is a proagent of Capsaicin and a first-in-class non-opioid TRPV1 agonist. It is designed to provide meaningful and long-lasting pain relief. This product is intended for research use only.
  • Proagent of capsaicin
  • First-in-class non-opioid TRPV1 agonist
  • Provides meaningful and long-lasting pain relief
  • Intended for research use only
Catalog No. 50-003-51790 Supplier Medchemexpress LLC Supplier No. HY137459A1MG
Vocacapsaicin hydrochloride is a proagent of Capsaicin and a first-in-class non-opioid TRPV1 agonist. It is designed to provide meaningful and long-lasting pain relief. This product is intended for research use only.
  • Proagent of capsaicin
  • First-in-class non-opioid TRPV1 agonist
  • Provides meaningful and long-lasting pain relief
  • Intended for research use only