Learn More
Medchemexpress LLC Vocacapsaicin hydrochloride | 1931116-92-5 | 95.7% | 496.08 | 1 ML
SDP
Catalog No. 5000349673
Encompass_Preferred
Vocacapsaicin hydrochloride (CA-008) is a proagent of Capsaicin and a first-in-class non-opioid TRPV1 agonist. Designed to provide meaningful and long-lasting pain relief, this product is intended for research use only.
  • Proagent of capsaicin
  • First-in-class non-opioid TRPV1 agonist
  • Provides meaningful and long-lasting pain relief
  • Involved in clinical trials for postsurgical pain
  • Acts as a TRP channel inhibitor
Catalog No. 50-003-49673 Supplier Medchemexpress LLC Supplier No. HY137459A10MM/1ML
Vocacapsaicin hydrochloride (CA-008) is a proagent of Capsaicin and a first-in-class non-opioid TRPV1 agonist. Designed to provide meaningful and long-lasting pain relief, this product is intended for research use only.
  • Proagent of capsaicin
  • First-in-class non-opioid TRPV1 agonist
  • Provides meaningful and long-lasting pain relief
  • Involved in clinical trials for postsurgical pain
  • Acts as a TRP channel inhibitor