Learn More
Medchemexpress LLC Vocacapsaicin hydrochloride | 1931116-92-5 | 95.7% | 496.08 g/mol | C26H42ClN3O4 | 5 MG
SDP
Catalog No. 5000222907
Encompass_Preferred
Vocacapsaicin hydrochloride is the hydrochloride salt of vocacapsaicin, a first-in-class, non-opioid TRPV1 agonist and prodrug of capsaicin. It is used in preclinical research to study TRPV1-mediated pain pathways and to evaluate long-lasting, local analgesic effects. Supplied as a purified powder for laboratory research use only.
  • Acts as a prodrug of capsaicin that targets TRPV1.
  • Provides prolonged local analgesic effects in preclinical models.
  • Non-opioid mechanism useful for pain pathway studies.
  • Hydrochloride salt form suitable for formulation and storage.
  • Available with documented purity for analytical reproducibility.
Catalog No. 50-002-22907 Supplier Medchemexpress LLC Supplier No. HY137459A5MG
Vocacapsaicin hydrochloride is the hydrochloride salt of vocacapsaicin, a first-in-class, non-opioid TRPV1 agonist and prodrug of capsaicin. It is used in preclinical research to study TRPV1-mediated pain pathways and to evaluate long-lasting, local analgesic effects. Supplied as a purified powder for laboratory research use only.
  • Acts as a prodrug of capsaicin that targets TRPV1.
  • Provides prolonged local analgesic effects in preclinical models.
  • Non-opioid mechanism useful for pain pathway studies.
  • Hydrochloride salt form suitable for formulation and storage.
  • Available with documented purity for analytical reproducibility.