Learn More
Medchemexpress LLC Vocacapsaicin hydrochloride | 1931116-92-5 | 95.7% | C26H42ClN3O4 | 10MG
SDP
Catalog No. 5000259651
Encompass_Preferred
Vocacapsaicin hydrochloride is a prodrug of capsaicin and a first-in-class, non-opioid TRPV1 agonist investigated for localized, long-lasting pain relief. Supplied as a solid research-grade compound, it is intended for in vitro and preclinical pharmacology and formulation studies.
  • Prodrug of capsaicin with TRPV1 agonist activity.
  • Provides localized, sustained analgesic effects in preclinical models.
  • High chemical purity suitable for research applications.
  • Soluble in DMSO at 100 mg/mL with ultrasonic assistance.
  • Recommended sealed storage to maintain stability.
Catalog No. 50-002-59651 Supplier Medchemexpress LLC Supplier No. HY137459A10MG
Vocacapsaicin hydrochloride is a prodrug of capsaicin and a first-in-class, non-opioid TRPV1 agonist investigated for localized, long-lasting pain relief. Supplied as a solid research-grade compound, it is intended for in vitro and preclinical pharmacology and formulation studies.
  • Prodrug of capsaicin with TRPV1 agonist activity.
  • Provides localized, sustained analgesic effects in preclinical models.
  • High chemical purity suitable for research applications.
  • Soluble in DMSO at 100 mg/mL with ultrasonic assistance.
  • Recommended sealed storage to maintain stability.