Learn More
Medchemexpress LLC Vociprotafib (RMC-4630) | 2172652-48-9 | 99.70% | C20H27ClN6O2S | 100 MG
SDP
Catalog No. 5000354815
Encompass_Preferred
Vociprotafib (RMC-4630) is an orally active, selective, and potent phosphatase SHP2 inhibitor. It functions by blocking the activation of the RAS-RAF-MEK-ERK signaling pathway, demonstrating antitumor activity. This compound accelerates tumor regressions in Osimertinib-sensitive EGFR-mutant tumors and delays tumor regrowth after treatment.
  • Orally active, selective, and potent phosphatase SHP2 inhibitor
  • Blocks activation of the RAS-RAF-MEK-ERK signaling pathway
  • Demonstrates antitumor activity
  • Accelerates tumor regressions in EGFR-mutant tumors
  • Delays tumor regrowth after treatment cessation
  • Effective against tumors resistant to Osimertinib via c-MET amplification
  • Investigated in a phase 1 clinical trial for KRAS-mutant non-small cell lung cancer
  • Considered a foundational approach for targeted therapy combinations in RAS pathway driven cancers
Catalog No. 50-003-54815 Supplier Medchemexpress LLC Supplier No. HY141523100MG
Vociprotafib (RMC-4630) is an orally active, selective, and potent phosphatase SHP2 inhibitor. It functions by blocking the activation of the RAS-RAF-MEK-ERK signaling pathway, demonstrating antitumor activity. This compound accelerates tumor regressions in Osimertinib-sensitive EGFR-mutant tumors and delays tumor regrowth after treatment.
  • Orally active, selective, and potent phosphatase SHP2 inhibitor
  • Blocks activation of the RAS-RAF-MEK-ERK signaling pathway
  • Demonstrates antitumor activity
  • Accelerates tumor regressions in EGFR-mutant tumors
  • Delays tumor regrowth after treatment cessation
  • Effective against tumors resistant to Osimertinib via c-MET amplification
  • Investigated in a phase 1 clinical trial for KRAS-mutant non-small cell lung cancer
  • Considered a foundational approach for targeted therapy combinations in RAS pathway driven cancers