Learn More
Medchemexpress LLC HY-12142 5mg Medchemexpress, Vofopitant CAS:168266-90-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY121425MG
Encompass_Preferred
Medchemexpress, HY-12142 5mg Vofopitant CAS:168266-90-8 Vofopitant is potent tachykinin NK 1 receptor antagonist, with pK i s of 10.6, 9.5, and 9.8 for human, rat and ferret NK 1 receptor, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2558