Learn More
Medchemexpress LLC Vofopitant dihydrochloride | 168266-51-1 | 99.6% | 1 ML
SDP
Catalog No. 5000232873
Encompass_Preferred
Vofopitant dihydrochloride is a selective tachykinin neurokinin 1 (NK1) receptor antagonist supplied as a research reagent. It is available as a solid or as a 10 mM solution in DMSO for in vitro pharmacology and biochemical assays. Not for human or diagnostic use.
  • Selective NK1 receptor antagonist for receptor binding and functional assays.
  • Available as solid or 10 mM solution in DMSO for convenient dosing.
  • High chemical purity suitable for preclinical research.
  • Storable at 4°C (solid) or long-term frozen in solvent to preserve stability.
Catalog No. 50-002-32873 Supplier Medchemexpress LLC Supplier No. HY1214310MM 1ML
Vofopitant dihydrochloride is a selective tachykinin neurokinin 1 (NK1) receptor antagonist supplied as a research reagent. It is available as a solid or as a 10 mM solution in DMSO for in vitro pharmacology and biochemical assays. Not for human or diagnostic use.
  • Selective NK1 receptor antagonist for receptor binding and functional assays.
  • Available as solid or 10 mM solution in DMSO for convenient dosing.
  • High chemical purity suitable for preclinical research.
  • Storable at 4°C (solid) or long-term frozen in solvent to preserve stability.