Learn More
TARGETMOL CHEMICALS INC Vofopitant dihydrochloride 1MG
SDP
Catalog No. 502879564
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Vofopitant dihydrochloride (GR 205171A) is an orally available, selective tachykinin NK1 receptor antagonist that inhibits [3H]SP binding to NK1 with pKi values of 9.5 and 10.6, respectively. It is a potential compound for the treatment of pathological vomiting. Purity 98.99%
Catalog No. 50-287-9564 Supplier TARGETMOL CHEMICALS INC Supplier No. T13329L1MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Vofopitant dihydrochloride (GR 205171A) is an orally available, selective tachykinin NK1 receptor antagonist that inhibits [3H]SP binding to NK1 with pKi values of 9.5 and 10.6, respectively. It is a potential compound for the treatment of pathological vomiting. Purity 98.99%