Learn More
NEOGEN Vogel Johnson Agar, 500 g
SDP
Catalog No. 502015936
Encompass_Preferred
Vogel Johnson Agar is used for the isolation of staphylococci in a laboratory setting. Vogel Johnson Agar is not intended for use in the diagnosis of disease or other conditions in humans. Coagulase-positive staphylococci, primarily Staphylococcus aureus, are among the microorganisms that cause spoilage or chemical changes in cosmetic products. To isolate coagulase-positive, mannitol fermenting staphylococci, Vogel and Johnson modified Tellurite-Glycine Agar by Zebovitz, Evans, and Niven. The modification included increasing mannitol and adding a pH indicator. Vogel Johnson Agar selects and differentiates coagulase-positive staphylococci that ferment mannitol and reduce tellurite. Vogel Johnson (VJ) Agar is specified in standard methods testing for cosmetics and nutritional supplements.For more information visit https://www.neogen.com/solutions/Microbiology/vogel-johnson-agar
Catalog No. 50-201-5936 Supplier NEOGEN Supplier No. 700003656
Vogel Johnson Agar is used for the isolation of staphylococci in a laboratory setting. Vogel Johnson Agar is not intended for use in the diagnosis of disease or other conditions in humans. Coagulase-positive staphylococci, primarily Staphylococcus aureus, are among the microorganisms that cause spoilage or chemical changes in cosmetic products. To isolate coagulase-positive, mannitol fermenting staphylococci, Vogel and Johnson modified Tellurite-Glycine Agar by Zebovitz, Evans, and Niven. The modification included increasing mannitol and adding a pH indicator. Vogel Johnson Agar selects and differentiates coagulase-positive staphylococci that ferment mannitol and reduce tellurite. Vogel Johnson (VJ) Agar is specified in standard methods testing for cosmetics and nutritional supplements.For more information visit https://www.neogen.com/solutions/Microbiology/vogel-johnson-agar