Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ Vogel and Johnson Agar
SDP

Catalog No. 23001783 Shop All Hardy Diagnostics Products
Change view
Click to view available options
No. per Pack:
10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
23-001-783 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-783 Supplier Hardy Diagnostics™ Supplier No. G193
Add to Cart
Add to Cart
This item is not returnable. View return policy

For the selective isolation of coagulase-positive, mannitol-positive Staphylococcus aureus strains in clinical specimens and foods.

  • Selective agents incorporated into the medium are glycine and lithium chloride, which inhibit the growth of most other microorganisms
  • Potassium tellurite is an inhibitory agent and is easily reduced by coagulase-positive staphylococci, leaving a black precipitate in the colony
  • The Association of Analytical Chemists (AOAC) has recommended the medium for detection of S. aureus in foods
  • The pH indicator phenol red enables the detection of mannitol fermentation, a differentiating characteristic of most strains of Staphylococcus aureus

Specifications

Appearance Slightly Opalescent
Certifications/Compliance ISO 13485
Color Reddish-orange
Content And Storage 2°C to 8°C
Country of Origin United States
Additive Glycine, Mannitol, Dipotassium Phosphate, Lithium Chloride, Phenol Red, Potassium Tellurite
Detects Coagulase-positive, Mannitol-positive Staphylococcus aureus Strains
Manufacturing Origin United States
Product Type Agar Plate
Target Organism Class Gram-positive Organisms
Test Type Microbial Detection
Fill Volume 19 mL
Formulation Pancreatic Digest of Casein - 10.0gm, Glycine - 10.0gm, Mannitol - 10.0gm, Yeast Extract - 5.0gm, Dipotassium Phosphate - 5.0gm, Lithium Chloride - 5.0gm, Phenol Red - 25.0mg, Agar - 15.0gm, Potassium Tellurite, 1% - 20.0mL
For Use With (Application) Detection of coagulase-positive, mannitol-positive Staphylococcus aureus strains in clinical specimens and foods
Incubation Conditions 35°C
Incubation Time 18 to 48 hr.
No. per Pack 10
Packaging Type Bag
Quantity 10 Plates
Dimensions (Dia. x H) 100 x 15 mm
Width (Metric) 100 mm
Height (Metric) 15 mm
Depth (Metric) 100 mm
Diameter (Metric) 100 mm
Species Staphylococcus species, Escherichia coli, Enterococcus faecalis
Protein Source Pancreatic Digest of Casein, Yeast Extract
Manufacturing Quality ISO 13485
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.