Learn More
Alkali Scientific Voges–Proskauer Reagent B (40% KOH), 25 mL Tube, 1/Pk
SDP
Supplier:  Alkali Scientific VPBR00
Encompass_Preferred
Voges–Proskauer Reagent B (40% KOH), 25 mL Tube, 1/Pk. This reagent is commonly used in biochemical testing, particularly for testing the presence of acetoin in bacterial cultures. The 40% potassium hydroxide (KOH) solution provides accurate and reliable results when conducting laboratory tests.
Catalog No. 50-006-62559