Learn More
Ace Glass, Inc. Volumetric flask, class A, 100mL, amber, white lettering, with individual certificate, includes 14/23 polyethylene stopper, sp/2
SDP
Catalog No. 5097487
Encompass
Volumetric flask, class A, 100mL, amber, white lettering, with individual certificate, includes 14/23 polyethylene stopper, sp/2
Catalog No. 50-974-87 Supplier Ace Glass, Inc. Supplier No. 712314
Volumetric flask, class A, 100mL, amber, white lettering, with individual certificate, includes 14/23 polyethylene stopper, sp/2