Learn More
Ace Glass, Inc. Volumetric flask, class A, 10mL, amber, white lettering, with individual certificate, includes 10/19 polyethylene stopper, sp/2
SDP
Catalog No. 5097483
Encompass
Volumetric flask, class A, 10mL, amber, white lettering, with individual certificate, includes 10/19 polyethylene stopper, sp/2
Catalog No. 50-974-83 Supplier Ace Glass, Inc. Supplier No. 712304
Volumetric flask, class A, 10mL, amber, white lettering, with individual certificate, includes 10/19 polyethylene stopper, sp/2