Learn More
Ace Glass, Inc. Volumetric flask, class A, 2000mL, amber, white lettering, with individual certificate, includes 29/32 polyethylene stopper, sp/2
SDP
Catalog No. 50979103
Encompass
2000mL Class A amber volumetric flask, used to precisely measure specified amounts of liquid in conjunction with the preparation and storage of solutions. These Class A flasks are in accordance with DIN and ISO accuracy standards to contain, and feature white lettering with the individual number and production date lasered onto its base. Each flask is tested individually, and the uncertainty measurement and date of issue are recorded on the Certificate. Flask sequential numbers are 3 letters & 4 numbers (AAA-0001). Includes 29/32 polyethylene stopper. Sold in packs of 2.
Catalog No. 50-979-103 Supplier Ace Glass, Inc. Supplier No. 712324
2000mL Class A amber volumetric flask, used to precisely measure specified amounts of liquid in conjunction with the preparation and storage of solutions. These Class A flasks are in accordance with DIN and ISO accuracy standards to contain, and feature white lettering with the individual number and production date lasered onto its base. Each flask is tested individually, and the uncertainty measurement and date of issue are recorded on the Certificate. Flask sequential numbers are 3 letters & 4 numbers (AAA-0001). Includes 29/32 polyethylene stopper. Sold in packs of 2.