Learn More
Ace Glass, Inc. Volumetric flask, class A, 5000mL, blue lettering, with individual certificate, includes 34/35 polyethylene stopper
SDP
Catalog No. 50979100
Encompass
Duran Class A, 5000mL volumetric flask. Flasks maintain DIN and ISO accuracy standards to contain and feature blue lettering with the individual number and production date lasered onto its base. Flask includes certificate and 34/35 polyethylene stopper.
Catalog No. 50-979-100 Supplier Ace Glass, Inc. Supplier No. 712232
Duran Class A, 5000mL volumetric flask. Flasks maintain DIN and ISO accuracy standards to contain and feature blue lettering with the individual number and production date lasered onto its base. Flask includes certificate and 34/35 polyethylene stopper.