Learn More
Ace Glass, Inc. Volumetric flask, class A, 500mL, amber, white lettering, with batch certificate, includes 19/26 polyethylene stopper, sp/2
SDP
Catalog No. 5097468
Encompass
Volumetric flask, class A, 500mL, amber, white lettering, with batch certificate, includes 19/26 polyethylene stopper, sp/2
Catalog No. 50-974-68 Supplier Ace Glass, Inc. Supplier No. 711920
Volumetric flask, class A, 500mL, amber, white lettering, with batch certificate, includes 19/26 polyethylene stopper, sp/2