Learn More
Ace Glass, Inc. Volumetric flask, class A, 50mL, amber, white lettering, with batch certificate, includes 12/21 polyethylene stopper, sp/2
SDP
Catalog No. 5097464
Encompass
Volumetric flask, class A, 50mL, amber, white lettering, with batch certificate, includes 12/21 polyethylene stopper, sp/2
Catalog No. 50-974-64 Supplier Ace Glass, Inc. Supplier No. 711910
Volumetric flask, class A, 50mL, amber, white lettering, with batch certificate, includes 12/21 polyethylene stopper, sp/2