Learn More
CHEMSCIENCE Pipette Volumetric, With One Mark, Accuracy As Per Class A, Capacity(mL) 6.5, Individually Serialized, Tolerance ±0.01 mL
Catalog No. NC2310941
Encompass_Preferred
Pipette Volumetric, With One Mark, Accuracy As Per Class A, Capacity(mL) 6.5, Individually Serialized, Tolerance ±0.01 mL
Catalog No. NC2310941 Supplier CHEMSCIENCE Supplier No. 123.302.06A
Pipette Volumetric, With One Mark, Accuracy As Per Class A, Capacity(mL) 6.5, Individually Serialized, Tolerance ±0.01 mL