Learn More
Andwin Scientific Pipet-Lite LTS Pipette L-1000XLS+
SDP
Supplier:  Andwin Scientific 17014382
Encompass_Preferred
Pipet-Lite XLS+ manual single-channel pipette, 100-1000 uL, uses LTS LiteTouch tips. Accurate and precise, ergonomic, autoclavable liquid end, rugged and durable (L-1000XLS+).
Catalog No. NC1552028