Learn More
Medchemexpress LLC Volanesorsen | 915430-78-3 | 98.0% | C230H320N63O125P19S19 | 1 MG
SDP
Catalog No. 5000355052
Encompass_Preferred
Volanesorsen (ISIS 304801) is an antisense oligonucleotide inhibitor of apolipoprotein CIII (apo-CIII) mRNA. It is known to reduce triglyceride levels and improve insulin resistance.
  • Reduces triglyceride levels
  • Improves insulin resistance
  • Being studied for hypertriglyceridemia, familial chylosiderosis syndrome, and type 2 diabetes
Catalog No. 50-003-55052 Supplier Medchemexpress LLC Supplier No. HY1457271MG
Volanesorsen (ISIS 304801) is an antisense oligonucleotide inhibitor of apolipoprotein CIII (apo-CIII) mRNA. It is known to reduce triglyceride levels and improve insulin resistance.
  • Reduces triglyceride levels
  • Improves insulin resistance
  • Being studied for hypertriglyceridemia, familial chylosiderosis syndrome, and type 2 diabetes