Learn More
Medchemexpress LLC HY-12137 10mg Medchemexpress, Volasertib CAS:755038-65-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1213710MG
Encompass_Preferred
Medchemexpress, HY-12137 10mg Volasertib CAS:755038-65-4 Volasertib is a highly potent Polo-like kinase 1 (PLK1) inhibitor with an IC50 of 0.87 nM, as well as the two closely related kinases PLK2 and PLK3 with IC50s of 5 and 56 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8855