Learn More
Hach Company Volatile Acid Standard Solution, 62,500 mg/L as Acetic Acid, pk/16 - 10 mL Voluette Ampules
Catalog No. 501994978 Shop All Hach Company Products
Encompass
Acetate Standard Solution. For accuracy checks in volatile acids determinations.
Catalog No. 50-199-4978 Supplier Hach Company Supplier No. 1427010
Acetate Standard Solution. For accuracy checks in volatile acids determinations.