Learn More
Perkin Elmer US LLC Volatile Capsule O-Ring Kit
Catalog No. 502112991
Encompass_Preferred
These high pressure pans and covers contain a sample in a sealed environment throughout an experiment. This ensures that the heat generated from vaporization or a volatile reaction does not interfere with the analyzer's measurements.
Catalog No. 50-211-2991 Supplier Perkin Elmer US LLC Supplier No. 03191535
These high pressure pans and covers contain a sample in a sealed environment throughout an experiment. This ensures that the heat generated from vaporization or a volatile reaction does not interfere with the analyzer's measurements.